Learn More
Bachem Fmoc-Cys(Acm)-OH
SDP
Supplier:  Bachem 4006622.0100
Encompass_Preferred
Fmoc-Cys(Acm)-OH, 86060-81-3, C21H22N2O5S, Mr 414.48, 100g. Following Fmoc-SPPS, Acm can be converted on resin into the S-carbomethoxysulfenyl (Scm) group. Scm is cleaved by reduction with DTT, treatment with thiols yields unsymmetrical disulfides.
Catalog No. 50-312-554