Learn More
Fmoc-Cys Bzl -OH, Advanced ChemTech
Specifications
Specifications
| CAS | 53298-33-2 |
| Color | White/Off-White |
| Molecular Weight (g/mol) | 433.06 |
| Quantity | 1 g |
| Grade | Biochemical |
| Chemical Name or Material | Fmoc-Cys Bzl -OH |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.