Learn More
Bachem Fmoc-Cys((RS)-2,3-di(palmitoyloxy)-propyl)-OH
SDP
Catalog No. 50258801
Encompass_Preferred
Fmoc-Cys((RS)-2,3-di(palmitoyloxy)-propyl)-OH, 210532-98-2, C53H83NO8S, Mr 894.31, 1g. Fmoc-Pam2Cys-OH. Incorporation of Fmoc-Pam2Cys-OH yields more hydrophilic products than incorporation of Pam3Cys-OH due to the free amino group obtained after attachment and deprotection. Nevertheless, the H-Pam2-peptides show comparable adjuvant activity.
Catalog No. 50-258-801 Supplier Bachem Supplier No. 4038236.1000
Fmoc-Cys((RS)-2,3-di(palmitoyloxy)-propyl)-OH, 210532-98-2, C53H83NO8S, Mr 894.31, 1g. Fmoc-Pam2Cys-OH. Incorporation of Fmoc-Pam2Cys-OH yields more hydrophilic products than incorporation of Pam3Cys-OH due to the free amino group obtained after attachment and deprotection. Nevertheless, the H-Pam2-peptides show comparable adjuvant activity.