Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Fmoc-Cys(Stbu)-Oh

Catalog No. 476311G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-631-1G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-631-1G Supplier Sigma Aldrich Supplier No. 476311G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Fmoc-Cys(Stbu)-Oh

Specifications

CAS 2964-48-9
Melting Point 163°C to 166°C
Molecular Formula C9H12N2O4
Linear Formula O2NC6H4CH(OH)CH(NH2)CH2OH
MDL Number MFCD00007359
Quantity 10 g
Synonym Nalpha-Fmoc-S-tert-butylthio-L-cysteine
Molecular Weight (g/mol) 212.2 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.