Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-Cys(Trt)-OH-1,2,3-13C3,15N | 360795-94-4 | 589.68 | 5 MG
SDP

Catalog No. 5000447069
Encompass_Preferred

Fmoc-Cys(Trt)-OH-1,2,3-13C3,15N is a 15N-labeled and 13C-labeled Fmoc-Cys(Trt)-OH-1,2,3, which is a cysteine derivative.

  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS

Catalog No. 50-004-47069 Supplier Medchemexpress LLC Supplier No. HYW007798S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Cys(Trt)-OH-1,2,3-13C3,15N is a 15N-labeled and 13C-labeled Fmoc-Cys(Trt)-OH-1,2,3, which is a cysteine derivative.

  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.