Learn More
Medchemexpress LLC Fmoc-Cys(Trt)-OH-d2 | 360565-11-3 | 587.72 | 5 MG
SDP
Catalog No. 5000448594
Encompass_Preferred
Fmoc-Cys(Trt)-OH-d2 is the deuterium labeled Fmoc-Cys(Trt)-OH, which is an N-terminal protected cysteine derivative.
  • Used as a tracer for quantitative analysis.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-48594 Supplier Medchemexpress LLC Supplier No. HYW007798S15MG
Add to Cart
Add to Cart
Fmoc-Cys(Trt)-OH-d2 is the deuterium labeled Fmoc-Cys(Trt)-OH, which is an N-terminal protected cysteine derivative.
  • Used as a tracer for quantitative analysis.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.