Learn More
Medchemexpress LLC Fmoc-Cys(Trt)-OH-d2 | 360565-11-3 | 587.72 | 50 MG
SDP
Catalog No. 5000446694
Encompass_Preferred
Fmoc-Cys(Trt)-OH-d2 is the deuterium labeled Fmoc-Cys(Trt)-OH, which is an N-terminal protected cysteine derivative.
  • Can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-46694 Supplier Medchemexpress LLC Supplier No. HYW007798S150MG
Fmoc-Cys(Trt)-OH-d2 is the deuterium labeled Fmoc-Cys(Trt)-OH, which is an N-terminal protected cysteine derivative.
  • Can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.