Learn More
Bachem Fmoc-cysteamine-2-methoxy-4-alkoxybenzyl alcohol resin
SDP
Catalog No. NC1820164
Encompass_Preferred
Fmoc-cysteamine-2-methoxy-4-alkoxybenzyl alcohol resin, 1g. Fmoc-2-aminoethanethiol-SASRINTM resin, Fmoc-2-mercaptoethylamine-SASRINTM resin. Convenient tool for the synthesis of b-thiol containing oligopeptides suitable for chemical conjugation or for the formation of organic monolayers on metal substrates. The cleavage from the resin with TFA/water/triisopropylsilane (94:4:2, v/v/v) yields C-terminal thiols. Linkage of these thiols e.g. to maleimides can be performed under very mild conditions.
Catalog No. NC1820164 Supplier Bachem Supplier No. 4026901.0001
Fmoc-cysteamine-2-methoxy-4-alkoxybenzyl alcohol resin, 1g. Fmoc-2-aminoethanethiol-SASRINTM resin, Fmoc-2-mercaptoethylamine-SASRINTM resin. Convenient tool for the synthesis of b-thiol containing oligopeptides suitable for chemical conjugation or for the formation of organic monolayers on metal substrates. The cleavage from the resin with TFA/water/triisopropylsilane (94:4:2, v/v/v) yields C-terminal thiols. Linkage of these thiols e.g. to maleimides can be performed under very mild conditions.