Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-D-2-Cyanophe | 25 g | CAS 401620-74-4 | MDL MFCD01863770

Catalog No. 502370181
Encompass

Fmoc-D-2-Cyanophe is a Amino Acid reagent (Subcategory: Phe) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 401620-74-4
- MDL: MFCD01863770
- InChIKey: ACFGBBASKOIPEW-HSZRJFAPSA-N
- Molecular Weight: 412.445
- Molecular Formula: C25H20N2O4
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29269070
- Country Of Origin: China
- IUPAC: (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-cyanophenyl)propanoic acid
- SMILES: N#CC1=CC=CC=C1C[C@H](C(O)=O)NC(OCC2C3=CC=CC=C3C4=CC=CC=C42)=O

Catalog No. 50-237-0181 Supplier STA Pharmaceutical Supplier No. STAAA1112625GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-D-2-Cyanophe is a Amino Acid reagent (Subcategory: Phe) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 401620-74-4
- MDL: MFCD01863770
- InChIKey: ACFGBBASKOIPEW-HSZRJFAPSA-N
- Molecular Weight: 412.445
- Molecular Formula: C25H20N2O4
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29269070
- Country Of Origin: China
- IUPAC: (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-cyanophenyl)propanoic acid
- SMILES: N#CC1=CC=CC=C1C[C@H](C(O)=O)NC(OCC2C3=CC=CC=C3C4=CC=CC=C42)=O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.