Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-D-2-Nitrophe | 50 g | CAS 478183-70-9 | MDL MFCD01317714

Catalog No. 502358556
Encompass

Fmoc-D-2-Nitrophe is a Amino Acid reagent (Subcategory: Phe) sold by WuXi TIDES. Offered in 50 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 478183-70-9
- MDL: MFCD01317714
- InChIKey: KVLRWXBYPKSBFY-OAQYLSRUSA-N
- Molecular Weight: 432.432
- Molecular Formula: C24H20N2O6
- Purity: ≥95%
- Container Type: 250 mL HDPE
- Pack Size: 50 g
- Net Weight: 50 g
- Gross Weight: 89.8 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-nitrophenyl)propanoic acid
- SMILES: O=C([C@@H](CC1=CC=CC=C1[N+]([O-])=O)NC(OCC2C3=CC=CC=C3C4=CC=CC=C42)=O)O

Catalog No. 50-235-8556 Supplier STA Pharmaceutical Supplier No. STAAA1113650GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-D-2-Nitrophe is a Amino Acid reagent (Subcategory: Phe) sold by WuXi TIDES. Offered in 50 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 478183-70-9
- MDL: MFCD01317714
- InChIKey: KVLRWXBYPKSBFY-OAQYLSRUSA-N
- Molecular Weight: 432.432
- Molecular Formula: C24H20N2O6
- Purity: ≥95%
- Container Type: 250 mL HDPE
- Pack Size: 50 g
- Net Weight: 50 g
- Gross Weight: 89.8 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-nitrophenyl)propanoic acid
- SMILES: O=C([C@@H](CC1=CC=CC=C1[N+]([O-])=O)NC(OCC2C3=CC=CC=C3C4=CC=CC=C42)=O)O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.