Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-D-Ala-OH | 79990-15-1 | MFCD00062960 | 311.33 | 500 G
SDP

Catalog No. 5000447366
Encompass_Preferred

Fmoc-D-Ala-OH is an alanine derivative intended for research use only. This compound is not for patient use and is recognized as a beneficial ergogenic dietary substance due to its influence on anabolic hormones and exercise-related performance.

  • Alanine derivative for research purposes
  • Affects secretion of anabolic hormones
  • Provides fuel during exercise
  • Enhances mental performance under stress
  • Helps prevent exercise-induced muscle damage
  • Molecular Formula: C18H17NO4
  • Appearance: White to off-white solid

Catalog No. 50-004-47366 Supplier Medchemexpress LLC Supplier No. HYW008072500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-D-Ala-OH is an alanine derivative intended for research use only. This compound is not for patient use and is recognized as a beneficial ergogenic dietary substance due to its influence on anabolic hormones and exercise-related performance.

  • Alanine derivative for research purposes
  • Affects secretion of anabolic hormones
  • Provides fuel during exercise
  • Enhances mental performance under stress
  • Helps prevent exercise-induced muscle damage
  • Molecular Formula: C18H17NO4
  • Appearance: White to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.