Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-D-Asp-OAll | 10 g | CAS 204246-17-3 | MDL MFCD01074689

Catalog No. 502344107
Encompass

Fmoc-D-Asp-OAll is a Amino Acid reagent (Subcategory: Asp) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 204246-17-3
- MDL: MFCD01074689
- InChIKey: ZJMVIWUCCRKNHY-LJQANCHMSA-N
- Molecular Weight: 395.411
- Molecular Formula: C22H21NO6
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (R)-3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-(allyloxy)-4-oxobutanoic acid
- SMILES: C=CCOC([C@@H](CC(O)=O)NC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)=O

Catalog No. 50-234-4107 Supplier STA Pharmaceutical Supplier No. STAAA1183410GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-D-Asp-OAll is a Amino Acid reagent (Subcategory: Asp) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 204246-17-3
- MDL: MFCD01074689
- InChIKey: ZJMVIWUCCRKNHY-LJQANCHMSA-N
- Molecular Weight: 395.411
- Molecular Formula: C22H21NO6
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (R)-3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-(allyloxy)-4-oxobutanoic acid
- SMILES: C=CCOC([C@@H](CC(O)=O)NC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)=O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.