Learn More
Medchemexpress LLC Fmoc-D-Cit-OH | 200344-33-8 | MFCD00151942 | 97% | 397.42 | 10 G
SDP
Catalog No. 5000351256
Encompass_Preferred
Fmoc-D-Cit-OH is citrulline with an Fmoc protecting group. It is utilized in the synthesis of bioactive peptide mimetics, such as H-Dmt-D-Cit-Aba-b-Ala-NMe-30,50-(CF3)2-Bn and H-Dmt-D-Cit-Aba-b-Ala-NMe-Bn, which exhibit neurokinin-1 antagonist activity.
  • Used for research purposes only
  • Utilized in the synthesis of bioactive peptide mimetics
  • Exhibits neurokinin-1 antagonist activity
Catalog No. 50-003-51256 Supplier Medchemexpress LLC Supplier No. HY13810610G
Fmoc-D-Cit-OH is citrulline with an Fmoc protecting group. It is utilized in the synthesis of bioactive peptide mimetics, such as H-Dmt-D-Cit-Aba-b-Ala-NMe-30,50-(CF3)2-Bn and H-Dmt-D-Cit-Aba-b-Ala-NMe-Bn, which exhibit neurokinin-1 antagonist activity.
  • Used for research purposes only
  • Utilized in the synthesis of bioactive peptide mimetics
  • Exhibits neurokinin-1 antagonist activity