Learn More
Bachem Fmoc-p-chloro-D-Phe-OH
SDP
Supplier:  Bachem 4015614.0005
Encompass_Preferred
Fmoc-p-chloro-D-Phe-OH, 142994-19-2, C24H20ClNO4, Mr 421.88, 5g. Fmoc-4-chloro-D-Phe-OH, (R)-3-(4-Chloro-phenyl)-2-(Fmoc-amino)-propionic acid. Fmoc-D-Cpa-OH, building block for GnRH antagonists.
Catalog No. 50-194-7261