Learn More
BLD PHARMATECH CO LIMITED Fmoc-D-Cys(Trt)-Wang Resin, 5g, Purity: 100-200 mesh, 1% DVB, 0.3-0.8 mmol/g
Catalog No. NC3965361
Encompass_Preferred
Fmoc-D-Cys(Trt)-Wang Resin, 5g, Purity: 100-200 mesh, 1% DVB, 0.3-0.8 mmol/g
Catalog No. NC3965361 Supplier BLD PHARMATECH CO LIMITED Supplier No. BD6290875G
Fmoc-D-Cys(Trt)-Wang Resin, 5g, Purity: 100-200 mesh, 1% DVB, 0.3-0.8 mmol/g