Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-D-Cys(Trt)-Opfp | 5 g | CAS 200395-72-8 | MDL MFCD00153375

Catalog No. 502357797
Encompass

Fmoc-D-Cys(Trt)-Opfp is a Amino Acid reagent (Subcategory: Cys) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 200395-72-8
- MDL: MFCD00153375
- InChIKey: FDAUCYDVXIPBDR-UUWRZZSWSA-N
- Molecular Weight: 751.768
- Molecular Formula: C43H30F5NO4S
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29309016
- Country Of Origin: China
- IUPAC: perfluorophenyl N-(((9H-fluoren-9-yl)methoxy)carbonyl)-S-trityl-D-cysteinate
- SMILES: O=C([C@@H](CSC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)NC(OCC4C5=CC=CC=C5C6=CC=CC=C64)=O)OC7=C(C(F)=C(C(F)=C7F)F)F

Catalog No. 50-235-7797 Supplier STA Pharmaceutical Supplier No. STAAA108175GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-D-Cys(Trt)-Opfp is a Amino Acid reagent (Subcategory: Cys) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 200395-72-8
- MDL: MFCD00153375
- InChIKey: FDAUCYDVXIPBDR-UUWRZZSWSA-N
- Molecular Weight: 751.768
- Molecular Formula: C43H30F5NO4S
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29309016
- Country Of Origin: China
- IUPAC: perfluorophenyl N-(((9H-fluoren-9-yl)methoxy)carbonyl)-S-trityl-D-cysteinate
- SMILES: O=C([C@@H](CSC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)NC(OCC4C5=CC=CC=C5C6=CC=CC=C64)=O)OC7=C(C(F)=C(C(F)=C7F)F)F

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.