Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-D-His(Boc)-OH, cyclohexylammonium salt | 100 g | CAS 210755-31-0 | MDL MFCD00270617

Catalog No. 502362051
Encompass

Fmoc-D-His(Boc)-OH, cyclohexylammonium salt is a Amino Acid reagent (Subcategory: His) sold by WuXi TIDES. Offered in 100 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 210755-31-0
- MDL: MFCD00270617
- InChIKey: UDEXYAHZNOKVIG-VZYDHVRKSA-N
- Molecular Weight: 576.694
- Molecular Formula: C32H40N4O6
- Purity: ≥95%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29332990
- Country Of Origin: China
- IUPAC: cyclohexanamine Na-(((9H-fluoren-9-yl)methoxy)carbonyl)-Nt-(tert-butoxycarbonyl)-D-histidinate
- SMILES: CC(C)(C)OC(N(C=N1)C=C1C[C@H](C(O)=O)NC(OCC2C3=C(C4=C2C=CC=C4)C=CC=C3)=O)=O.NC5CCCCC5

Catalog No. 50-236-2051 Supplier STA Pharmaceutical Supplier No. STAAA11391100GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-D-His(Boc)-OH, cyclohexylammonium salt is a Amino Acid reagent (Subcategory: His) sold by WuXi TIDES. Offered in 100 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 210755-31-0
- MDL: MFCD00270617
- InChIKey: UDEXYAHZNOKVIG-VZYDHVRKSA-N
- Molecular Weight: 576.694
- Molecular Formula: C32H40N4O6
- Purity: ≥95%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29332990
- Country Of Origin: China
- IUPAC: cyclohexanamine Na-(((9H-fluoren-9-yl)methoxy)carbonyl)-Nt-(tert-butoxycarbonyl)-D-histidinate
- SMILES: CC(C)(C)OC(N(C=N1)C=C1C[C@H](C(O)=O)NC(OCC2C3=C(C4=C2C=CC=C4)C=CC=C3)=O)=O.NC5CCCCC5

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.