Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-D-Lys Aloc -OH, Advanced ChemTech

Catalog No. 01340230
missing translation for 'changeView'
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-340-230 5 g
01-340-231 25 g
01-340-232 1 g
01-340-233 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
Catalog No. 01-340-230 Supplier Advanced Chem Tech missing translation for 'supplierNo' FK33985G
Only null left
Add to cart
Add to cart

Chemical Identifiers

CAS 214750-75-1
Molecular Formula C25H28N2O6
Molecular Weight (g/mol) 452.51
MDL Number MFCD00798639
InChI Key OJBNDXHENJDCBA-JOCHJYFZSA-N
IUPAC Name (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-6-{[(prop-2-en-1-yloxy)carbonyl]amino}hexanoic acid
SMILES OC(=O)[C@@H](CCCCNC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Color White/Off-White
Quantity 5 g
Grade Biochemical
Chemical Name or Material Fmoc-D-Lys Aloc -OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.