Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-D-Lys Biotin -OH, Advanced ChemTech

Catalog No. 01340242
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-340-242 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-340-242 Supplier Advanced Chem Tech Supplier No. FK33861G
Add to Cart
Add to Cart

Fmoc-protected amino acid derivative for solid-phase peptide synthesis.

Chemical Identifiers

CAS 110990-09-5
Molecular Formula C31H38N4O6S
Molecular Weight (g/mol) 594.73
MDL Number MFCD00270742
InChI Key OFIBQNGDYNGUEZ-KAHNYGAISA-N
IUPAC Name (2R)-6-{5-[(3aS,4S,6aR)-2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanamido}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
SMILES [H][C@]12CS[C@@H](CCCCC(=O)NCCCC[C@@H](NC(=O)OCC3C4=C(C=CC=C4)C4=C3C=CC=C4)C(O)=O)[C@@]1([H])NC(=O)N2

Specifications

Color White/Off-White
Quantity 1 g
Grade Biochemical
Chemical Name or Material Fmoc-D-Lys Biotin -OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.