Learn More
Medchemexpress LLC Fmoc-D-Lys(N3)-OH | 1198791-53-5 | C21H22N4O4 | 100MG
SDP
Catalog No. 5000372075
Encompass_Preferred
Fmoc-D-Lys(N3)-OH is a click chemistry reagent that contains an azide group. It is suitable for the research of various biochemical applications, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules and ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • Click chemistry reagent
  • Contains an azide group
  • Used for research of various biochemical applications
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Enables ring strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-72075 Supplier Medchemexpress LLC Supplier No. HY151660100MG
Fmoc-D-Lys(N3)-OH is a click chemistry reagent that contains an azide group. It is suitable for the research of various biochemical applications, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules and ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • Click chemistry reagent
  • Contains an azide group
  • Used for research of various biochemical applications
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Enables ring strain-promoted alkyne-azide cycloaddition (SPAAC)