Learn More
Apexbio Technology LLC Fmoc-D-Lys(Boc)-OH 92122-45-7 5g
SDP
Catalog No. 502240410
Encompass_Preferred
Fmoc-D-Lys Boc -OH is an Fmoc-protected derivative of D-lysine with the side-chain amine group protected by a tert-butyloxycarbonyl (Boc) group It is primarily used in solid-phase peptide synthesis to facilitate the controlled incorporation of D-lysine residues into peptide chains thereby enabling the construction of peptides and cyclic peptides with defined stereochemistry Based on these properties Fmoc-D-Lys Boc -OH holds research potential in studies focusing on peptide receptor interactions cellular signaling pathways enzyme mechanisms and pharmaceutical development in biomedical sciences
This item is not returnable. View return policy
Catalog No. 50-224-0410 Supplier Apexbio Technology LLC Supplier No. A79355000
Fmoc-D-Lys Boc -OH is an Fmoc-protected derivative of D-lysine with the side-chain amine group protected by a tert-butyloxycarbonyl (Boc) group It is primarily used in solid-phase peptide synthesis to facilitate the controlled incorporation of D-lysine residues into peptide chains thereby enabling the construction of peptides and cyclic peptides with defined stereochemistry Based on these properties Fmoc-D-Lys Boc -OH holds research potential in studies focusing on peptide receptor interactions cellular signaling pathways enzyme mechanisms and pharmaceutical development in biomedical sciences