Learn More
Medchemexpress LLC Fmoc-D-Nle-OH | 112883-41-7 | 353.41 | 25 G
SDP
Catalog No. 5000449914
Encompass_Preferred
Fmoc-D-Nle-OH is a leucine derivative primarily for research use. This compound is recognized as a beneficial ergogenic dietary substance, capable of influencing anabolic hormone secretion and providing fuel during exercise.
  • Leucine derivative
  • Acts as an ergogenic supplement
  • Influences the secretion of anabolic hormones
  • Supplies fuel during exercise
  • Improves mental performance during stress-related tasks
  • Prevents exercise-induced muscle damage
  • For research use only
Catalog No. 50-004-49914 Supplier Medchemexpress LLC Supplier No. 25GHYW008273
Fmoc-D-Nle-OH is a leucine derivative primarily for research use. This compound is recognized as a beneficial ergogenic dietary substance, capable of influencing anabolic hormone secretion and providing fuel during exercise.
  • Leucine derivative
  • Acts as an ergogenic supplement
  • Influences the secretion of anabolic hormones
  • Supplies fuel during exercise
  • Improves mental performance during stress-related tasks
  • Prevents exercise-induced muscle damage
  • For research use only