Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-D-Nle-OH, Advanced ChemTech

Catalog No. 01340180
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Catalog No. Quantity
01-340-180 25 g
01-340-179 5 g
01-340-181 1 g
01-340-182 100 g
4 options

Catalog No. 01-340-180

Supplier: Advanced Chem Tech FL337125G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 112883-41-7
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.42
MDL Number MFCD00155639
InChI Key VCFCFPNRQDANPN-TWYLJJHKNA-N
IUPAC Name (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
SMILES CCCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Color White to Off-White
Quantity 25 g
Grade Biochemical
Chemical Name or Material Fmoc-D-Nle-OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.