Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-D-Oic-OH(2R,3aS,7aS) | 25 g | CAS 1217512-55-4 | MDL MFCD12198183

Catalog No. 502353408
Encompass

Fmoc-D-Oic-OH(2R,3aS,7aS) is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1217512-55-4
- MDL: MFCD12198183
- InChIKey: JBZXLQHJZHITMW-IVKVDSSUSA-N
- Molecular Weight: 391.467
- Molecular Formula: C24H25NO4
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29339980
- Country Of Origin: China
- IUPAC: (2R)-1-(((9H-fluoren-9-yl)methoxy)carbonyl)octahydro-1H-indole-2-carboxylic acid
- SMILES: O=C(O)[C@H](N1C(OCC2C3=CC=CC=C3C4=CC=CC=C42)=O)CC5C1CCCC5

Catalog No. 50-235-3408 Supplier STA Pharmaceutical Supplier No. STAAA1050025GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-D-Oic-OH(2R,3aS,7aS) is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1217512-55-4
- MDL: MFCD12198183
- InChIKey: JBZXLQHJZHITMW-IVKVDSSUSA-N
- Molecular Weight: 391.467
- Molecular Formula: C24H25NO4
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29339980
- Country Of Origin: China
- IUPAC: (2R)-1-(((9H-fluoren-9-yl)methoxy)carbonyl)octahydro-1H-indole-2-carboxylic acid
- SMILES: O=C(O)[C@H](N1C(OCC2C3=CC=CC=C3C4=CC=CC=C42)=O)CC5C1CCCC5

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.