Learn More
Medchemexpress LLC Fmoc-D-Phe(2,4-Cl2)-OH | 352351-61-2 | MFCD01863047 | 95.0% | 456.32 | C24H19Cl2NO4 | 100 MG
SDP
Catalog No. 5000283449
Encompass_Preferred
Fmoc-D-Phe(2,4-Cl2)-OH is an Fmoc-protected D-phenylalanine derivative carrying a 2,4-dichloro substitution. Supplied as a white to off-white solid, it is intended for use as a protected amino acid building block in solid-phase peptide synthesis and other organic synthesis applications where a dichloro-phenylalanine residue is required.
  • Fmoc-protected D-amino acid suitable for solid-phase peptide synthesis.
  • 2,4-dichloro substitution provides a modified aromatic side chain for SAR studies.
  • White to off-white solid, convenient to weigh and handle.
  • Stable under recommended storage conditions for extended shelf life.
  • Available in multiple package sizes, including 100 mg and larger quantities.
Catalog No. 50-002-83449 Supplier Medchemexpress LLC Supplier No. HYW141817100MG
Fmoc-D-Phe(2,4-Cl2)-OH is an Fmoc-protected D-phenylalanine derivative carrying a 2,4-dichloro substitution. Supplied as a white to off-white solid, it is intended for use as a protected amino acid building block in solid-phase peptide synthesis and other organic synthesis applications where a dichloro-phenylalanine residue is required.
  • Fmoc-protected D-amino acid suitable for solid-phase peptide synthesis.
  • 2,4-dichloro substitution provides a modified aromatic side chain for SAR studies.
  • White to off-white solid, convenient to weigh and handle.
  • Stable under recommended storage conditions for extended shelf life.
  • Available in multiple package sizes, including 100 mg and larger quantities.