Learn More
AnaSpec Fmoc-D-propargylglycine[Fmoc-D-Pra-OH] | MW 335.4 | M.F. C20H17NO4 (1 g)
Catalog No. 50351102
Encompass
Fmoc-D-propargylglycine[Fmoc-D-Pra-OH] ; MW 335.4; M.F. C20H17NO4 (1 g) An unusual amino acid used in Solid Phase Peptide Synthesis (SPPS) as: A) a precursor to the tritium-induced Norvaline peptides1, B) in the synthesis of branched peptides using a palladium-catalyzed coupling reaction2, and C) as a reagent in the dual-catalyst Sonogashira coupling reaction.3
Catalog No. 50351102 Supplier AnaSpec Supplier No. AS26265F1
Fmoc-D-propargylglycine[Fmoc-D-Pra-OH] ; MW 335.4; M.F. C20H17NO4 (1 g) An unusual amino acid used in Solid Phase Peptide Synthesis (SPPS) as: A) a precursor to the tritium-induced Norvaline peptides1, B) in the synthesis of branched peptides using a palladium-catalyzed coupling reaction2, and C) as a reagent in the dual-catalyst Sonogashira coupling reaction.3