Learn More
MilliporeSigma FMOC-D-SER(TBU)-OH
Catalog No. 502302871
Encompass
Fmoc-D-Ser(tBu)-OH; > /=98% by HPLC; White-light yellowish solid powder; EMD; Novabiochem; 5g; N-alpha-Fmoc-O-tert-butyl-D-serine; F.W. 383.4; C22H25NO5; M.P. 120 to 130 deg.C (248 to 266 deg.F
Catalog No. 50-230-2871 Supplier MilliporeSigma Supplier No. 8521560005
Fmoc-D-Ser(tBu)-OH; > /=98% by HPLC; White-light yellowish solid powder; EMD; Novabiochem; 5g; N-alpha-Fmoc-O-tert-butyl-D-serine; F.W. 383.4; C22H25NO5; M.P. 120 to 130 deg.C (248 to 266 deg.F