Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-D-Thr-OH H2O | 50 g | CAS 1272755-74-4 | MDL MFCD07366860

Catalog No. 502352633
Encompass

Fmoc-D-Thr-OH H2O is a Amino Acid reagent (Subcategory: Thr) sold by WuXi TIDES. Offered in 50 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1272755-74-4
- MDL: MFCD07366860
- InChIKey: CGNQPFAECJFQNV-VFZPIINCSA-N
- Molecular Weight: 359.378
- Molecular Formula: C19H21NO6
- Purity: ≥95%
- Container Type: 250 mL HDPE
- Pack Size: 50 g
- Net Weight: 50 g
- Gross Weight: 89.8 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (((9H-fluoren-9-yl)methoxy)carbonyl)-D-threonine hydrate
- SMILES: C[C@@H]([C@H](C(O)=O)NC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)O.O

Catalog No. 50-235-2633 Supplier STA Pharmaceutical Supplier No. STAAA1054750GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-D-Thr-OH H2O is a Amino Acid reagent (Subcategory: Thr) sold by WuXi TIDES. Offered in 50 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1272755-74-4
- MDL: MFCD07366860
- InChIKey: CGNQPFAECJFQNV-VFZPIINCSA-N
- Molecular Weight: 359.378
- Molecular Formula: C19H21NO6
- Purity: ≥95%
- Container Type: 250 mL HDPE
- Pack Size: 50 g
- Net Weight: 50 g
- Gross Weight: 89.8 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (((9H-fluoren-9-yl)methoxy)carbonyl)-D-threonine hydrate
- SMILES: C[C@@H]([C@H](C(O)=O)NC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)O.O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.