Learn More
Medchemexpress LLC Fmoc-D-Tic-OH | 130309-33-0 | 99.9% | 399.44 | 5 G
SDP
Catalog No. 5000454340
Encompass_Preferred
Fmoc-D-Tic-OH is an amino acid derivative with an Fmoc protecting group, which can be used to synthesize bioactive peptide mimetics, such as [desArg10]HOE 140, which has bradykinin B1 antagonist activity. It is for research use only and not sold to patients.
  • Amino acid derivative with an Fmoc protecting group.
  • Can be used to synthesize bioactive peptide mimetics.
  • Applicable in the synthesis of [desArg10]HOE 140, which has bradykinin B1 antagonist activity.
Catalog No. 50-004-54340 Supplier Medchemexpress LLC Supplier No. HYW0131535G
Fmoc-D-Tic-OH is an amino acid derivative with an Fmoc protecting group, which can be used to synthesize bioactive peptide mimetics, such as [desArg10]HOE 140, which has bradykinin B1 antagonist activity. It is for research use only and not sold to patients.
  • Amino acid derivative with an Fmoc protecting group.
  • Can be used to synthesize bioactive peptide mimetics.
  • Applicable in the synthesis of [desArg10]HOE 140, which has bradykinin B1 antagonist activity.