Learn More
Medchemexpress LLC HY-79129 5g Medchemexpress, Fmoc-D-Trp(Boc)-OH CAS:163619-04-3 Purity:>98%
SDP
Catalog No. 502082294
Encompass_Preferred
Medchemexpress, HY-79129 5g Fmoc-D-Trp(Boc)-OH CAS:163619-04-3 Fmoc-D-Trp(Boc)-OH is a cleavable ADC linker that used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2294 Supplier Medchemexpress LLC Supplier No. HY791295G
Medchemexpress, HY-79129 5g Fmoc-D-Trp(Boc)-OH CAS:163619-04-3 Fmoc-D-Trp(Boc)-OH is a cleavable ADC linker that used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.