Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-D-Tyr But -OH, Advanced ChemTech

Catalog No. 01339743
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-339-743 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-339-743 Supplier Advanced Chem Tech Supplier No. FY356325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 118488-18-9 | C28H29NO5 | 459.54 g/mol

Chemical Identifiers

CAS 118488-18-9
Molecular Formula C28H29NO5
Molecular Weight (g/mol) 459.54
MDL Number MFCD00065684
InChI Key JAUKCFULLJFBFN-KSYWNVGFNA-N
IUPAC Name (2R)-3-[4-(tert-butoxy)phenyl]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
SMILES CC(C)(C)OC1=CC=C(C[C@@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1

Specifications

Color White to Off-White
Quantity 25 g
Grade Biochemical
Chemical Name or Material Fmoc-D-Tyr But -OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.