Learn More
Bachem Fmoc-[D4]Ala-OH
SDP
Catalog No. 501947099
Encompass_Preferred
Fmoc-[D4]Ala-OH, 225101-69-9, C18H13D4NO4, Mr 315.37, 0.5g. Fmoc-(D4)-Ala-OH was used by Bowman and Zaia in the SPPS of peptide tags for the stable isotopic labeling of carbohydrates which allowed their mass spectroscopic quantification. For 2H-NMR-studies on the influence of Pro on the folding of WALP transmembrane peptides, Koeppe’s group incorporated pairs of deuterated alanines by introducing Fmoc-(D4)-Ala-OH (60-100%) at selected sequence positions.
Catalog No. 50-194-7099 Supplier Bachem Supplier No. 4071077.0500
Fmoc-[D4]Ala-OH, 225101-69-9, C18H13D4NO4, Mr 315.37, 0.5g. Fmoc-(D4)-Ala-OH was used by Bowman and Zaia in the SPPS of peptide tags for the stable isotopic labeling of carbohydrates which allowed their mass spectroscopic quantification. For 2H-NMR-studies on the influence of Pro on the folding of WALP transmembrane peptides, Koeppe’s group incorporated pairs of deuterated alanines by introducing Fmoc-(D4)-Ala-OH (60-100%) at selected sequence positions.