Learn More
Bachem Fmoc-Dap(ivDde)-OH
SDP
Supplier:  Bachem 4054323.0001
Encompass_Preferred
Fmoc-Dap(ivDde)-OH, 607366-20-1, C31H36N2O6, Mr 532.64, 1g. The ivDde protective group possesses higher orthogonality relatively to Fmoc in comparison to Dde.
Catalog No. 50-258-808