Learn More
Medchemexpress LLC Fmoc-Dha-OH | 261522-33-2 | MFCD09263343 | 99.6% | 309.32 g/mol | C18H15NO4 | 10 MG
SDP
Catalog No. 5000280928
Encompass_Preferred
Fmoc-Dha-OH is an Fmoc-protected dehydroalanine (Dha) amino acid derivative provided for research use in peptide synthesis and chemical biology. It is designed to enable incorporation of dehydroalanine residues during solid-phase peptide synthesis and to support downstream chemical modifications in biological research applications.
  • Fmoc-protected dehydroalanine suitable for peptide synthesis.
  • High purity reported by the manufacturer (99.63%).
  • Available in milligram-scale pack sizes for small-scale research use.
  • Compatible with standard solid-phase peptide synthesis protocols.
  • Enables introduction of dehydroalanine residues for further chemical modifications.
Catalog No. 50-002-80928 Supplier Medchemexpress LLC Supplier No. HYW34889610MG
Fmoc-Dha-OH is an Fmoc-protected dehydroalanine (Dha) amino acid derivative provided for research use in peptide synthesis and chemical biology. It is designed to enable incorporation of dehydroalanine residues during solid-phase peptide synthesis and to support downstream chemical modifications in biological research applications.
  • Fmoc-protected dehydroalanine suitable for peptide synthesis.
  • High purity reported by the manufacturer (99.63%).
  • Available in milligram-scale pack sizes for small-scale research use.
  • Compatible with standard solid-phase peptide synthesis protocols.
  • Enables introduction of dehydroalanine residues for further chemical modifications.