Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical Fmoc-DL-beta-Leu | 5 g | CAS 282524-93-0 | MDL MFCD01863209

Catalog No. 502351673
Encompass

Fmoc-DL-beta-Leu is a Amino Acid reagent (Subcategory: Beta AA) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 282524-93-0
- MDL: MFCD01863209
- InChIKey: KFDMTAMWOIQTOB-UHFFFAOYSA-N
- Molecular Weight: 353.418
- Molecular Formula: C21H23NO4
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-methylpentanoic acid
- SMILES: CC(C)C(CC(O)=O)NC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O

Catalog No. 50-235-1673 Supplier STA Pharmaceutical Supplier No. STAAA101525GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-DL-beta-Leu is a Amino Acid reagent (Subcategory: Beta AA) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 282524-93-0
- MDL: MFCD01863209
- InChIKey: KFDMTAMWOIQTOB-UHFFFAOYSA-N
- Molecular Weight: 353.418
- Molecular Formula: C21H23NO4
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4-methylpentanoic acid
- SMILES: CC(C)C(CC(O)=O)NC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.