Learn More
MilliporeSigma FMOC-DPR(BOC)-OH
Catalog No. 502301900
Encompass
Fmoc-Dpr(Boc)-OH; > /=95% by TLC and HPLC; Solid; EMD; Novabiochem; 5g; N-alpha-Fmoc-N-beta-tert-Boc-L-diaminopropionic acid; F.W. 426.5; C23H26N2O6; CAS: 162558-25-0
Catalog No. 50-230-1900 Supplier MilliporeSigma Supplier No. 8520870005
Fmoc-Dpr(Boc)-OH; > /=95% by TLC and HPLC; Solid; EMD; Novabiochem; 5g; N-alpha-Fmoc-N-beta-tert-Boc-L-diaminopropionic acid; F.W. 426.5; C23H26N2O6; CAS: 162558-25-0