Learn More
Bachem Fmoc-e-azido-Nle-OH
SDP
Supplier:  Bachem 4072394.1000
Encompass_Preferred
Fmoc-e-azido-Nle-OH, 159610-89-6, C21H22N4O4, Mr 394.43, 1g. Fmoc-Lys(N2)-OH. Lysine-derived building block for "click chemistry" and other reactions involving azides. The azido moiety of Lys(N3) may also be selectively converted into an amino group with triphenylphosphine in the presence of water.
Catalog No. 50-258-820