Learn More
MilliporeSigma FMOC-G-ABU-OH
Catalog No. 502301758
Encompass
Fmoc-Gamma-Abu-OH; > /=98% by TLC and HPLC; White to light yellowish solid powder; EMD; Novabiochem; 25g; Fmoc-GABA; F.W. 325.4; C19H19NO4; M.P. 170-180 deg.C (338-356 deg.F); CAS: 116821-47-7
Catalog No. 50-230-1758 Supplier MilliporeSigma Supplier No. 8520430025
Fmoc-Gamma-Abu-OH; > /=98% by TLC and HPLC; White to light yellowish solid powder; EMD; Novabiochem; 25g; Fmoc-GABA; F.W. 325.4; C19H19NO4; M.P. 170-180 deg.C (338-356 deg.F); CAS: 116821-47-7