Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fmoc-Gla OBut 2, Advanced ChemTech

Catalog No. 01340809
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-340-809 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-340-809 Supplier Advanced Chem Tech Supplier No. CB7002500MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 111662-64-7
Molecular Formula C29H35NO8
Molecular Weight (g/mol) 525.60
MDL Number MFCD00672341
InChI Key XJRUHYXXHXECDI-QHCPKHFHSA-N
IUPAC Name (2S)-5-(tert-butoxy)-4-[(tert-butoxy)carbonyl]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)C(C[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O)C(=O)OC(C)(C)C

Specifications

Color White/Off-White
Quantity 500 mg
Grade Biochemical
Chemical Name or Material Fmoc-Gla OBut 2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.