Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-Glu-OAll | 144120-54-7 | 409.43 | 10 G
SDP

Catalog No. 5000447307
Encompass_Preferred

Fmoc-Glu-OAll is a glutamic acid derivative. This compound is recognized as a beneficial ergogenic dietary substance, influencing anabolic hormone secretion, supporting fuel supply during exercise, enhancing mental performance under stress, and helping to prevent exercise-induced muscle damage.

  • Glutamic acid derivative
  • Influences anabolic hormone secretion
  • Supports fuel supply during exercise
  • Enhances mental performance under stress
  • Helps prevent exercise-induced muscle damage
  • White to off-white solid appearance
  • Molecular weight: 409.43
  • Purity: 98.9%

Catalog No. 50-004-47307 Supplier Medchemexpress LLC Supplier No. HYW00847510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Glu-OAll is a glutamic acid derivative. This compound is recognized as a beneficial ergogenic dietary substance, influencing anabolic hormone secretion, supporting fuel supply during exercise, enhancing mental performance under stress, and helping to prevent exercise-induced muscle damage.

  • Glutamic acid derivative
  • Influences anabolic hormone secretion
  • Supports fuel supply during exercise
  • Enhances mental performance under stress
  • Helps prevent exercise-induced muscle damage
  • White to off-white solid appearance
  • Molecular weight: 409.43
  • Purity: 98.9%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.