Learn More
MilliporeSigma FMOC-GLU(OALL)-OH
Supplier:  MilliporeSigma 8521230025
Encompass
Fmoc-Glu(OAll)-OH; > /=99.50% by HPLC; EMD; Novabiochem; 25g; F.W. 409.43; C23H23NO6; CAS: 133464-46-7
Catalog No. 50-230-1985