Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-Glu-OH | 121343-82-6 | 99.4% | 369.37 | 500 G
SDP

Catalog No. 5000366290
Encompass_Preferred

Fmoc-Glu-OH is a glutamic acid derivative. Amino acids and amino acid derivatives have been commercially used as ergogenic supplements. They influence the secretion of anabolic hormones, supply of fuel during exercise, mental performance during stress-related tasks, and prevent exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years. In solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility: DMSO: 100 mg/mL (270.73 mM; Needs ultrasonic; Hygroscopic DMSO has a significant impact on the solubility of the product, so newly opened DMSO should be used)

Catalog No. 50-003-66290 Supplier Medchemexpress LLC Supplier No. HYW011081500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Glu-OH is a glutamic acid derivative. Amino acids and amino acid derivatives have been commercially used as ergogenic supplements. They influence the secretion of anabolic hormones, supply of fuel during exercise, mental performance during stress-related tasks, and prevent exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years. In solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility: DMSO: 100 mg/mL (270.73 mM; Needs ultrasonic; Hygroscopic DMSO has a significant impact on the solubility of the product, so newly opened DMSO should be used)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.