Learn More
Medchemexpress LLC Fmoc-Glu-OMe | 145038-49-9 | MFCD22200617 | 99.7% | 383.40 g/mol | C21H21NO6 | 100 G
SDP
Catalog No. 5000209397
Encompass_Preferred
Fmoc-Glu-OMe is a Fmoc-protected glutamic acid methyl ester used as an amino acid derivative in peptide synthesis and biomaterials research. It has reported antibacterial activity and can form gels in silver nitrate (AgNO3) solutions, and has been investigated as a mouldable wound-healing biomaterial.
  • Fmoc-protected glutamic acid methyl ester
  • Reported antibacterial activity
  • Forms gels in AgNO3 solutions
  • Powder, typically stored at -20°C
  • High purity (reported ~99.74%)
  • Molecular weight 383.40 g/mol; formula C21H21NO6
Catalog No. 50-002-09397 Supplier Medchemexpress LLC Supplier No. HYW041988100G
Fmoc-Glu-OMe is a Fmoc-protected glutamic acid methyl ester used as an amino acid derivative in peptide synthesis and biomaterials research. It has reported antibacterial activity and can form gels in silver nitrate (AgNO3) solutions, and has been investigated as a mouldable wound-healing biomaterial.
  • Fmoc-protected glutamic acid methyl ester
  • Reported antibacterial activity
  • Forms gels in AgNO3 solutions
  • Powder, typically stored at -20°C
  • High purity (reported ~99.74%)
  • Molecular weight 383.40 g/mol; formula C21H21NO6