Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fmoc-Glu(OMe)-OH | 145038-50-2 | 383.39 | 25 G
SDP

Catalog No. 5000448258
Encompass_Preferred

Fmoc-Glu(OMe)-OH is a glutamic acid derivative. Amino acids and their derivatives are utilized commercially as ergogenic supplements, influencing the secretion of anabolic hormones, fuel supply during exercise, mental performance under stress, and preventing exercise-induced muscle damage. They are recognized for their benefits as ergogenic dietary substances.

  • Appearance: Solid
  • Color: White to off-white
  • Classification: Amino acid derivatives, Reagents for peptide synthesis, Amino acids and derivatives
  • Formula: C21H21NO6

Catalog No. 50-004-48258 Supplier Medchemexpress LLC Supplier No. 25GHYW008256
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Glu(OMe)-OH is a glutamic acid derivative. Amino acids and their derivatives are utilized commercially as ergogenic supplements, influencing the secretion of anabolic hormones, fuel supply during exercise, mental performance under stress, and preventing exercise-induced muscle damage. They are recognized for their benefits as ergogenic dietary substances.

  • Appearance: Solid
  • Color: White to off-white
  • Classification: Amino acid derivatives, Reagents for peptide synthesis, Amino acids and derivatives
  • Formula: C21H21NO6

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.