Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Fmoc-Glu(Otbu)-Oh

Catalog No. 4762525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-625-25G 25 g
47-625-1KG 1 kg
47-625-100G 100 g
47-625-10G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 47-625-25G Supplier Sigma Aldrich Supplier No. 4762525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Fmoc-Glu(OtBu)-OH (Fmoc-L-glutamic acid 5-tert-butyl ester) can be employed as a: Ligand in the synthesis of cis-substituted cyclopropane carboxylic acids via C-H activation of cyclopropane carboxamides using Pd catalyst.Linker in the preparation of multi-small molecule-conjugated PTX (paclitaxel) derivatives.It can be also used as a building block in the preparation of stapled +a-helical peptides , and peptide C-terminal thioesters .

Specifications

CAS 71989-18-9
Molecular Formula C24H27NO6
Linear Formula C24H27NO6
MDL Number MFCD00037135
Quantity 25 g
Synonym Fmoc-L-glutamic acid 5-tert-butyl ester
Molecular Weight (g/mol) 425.47 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.