Learn More
AnaSpec FMOC-GLU(OTBU)-OH
Catalog No. 50845555
Encompass
Fmoc-Glu(OtBu)-OH[N-a -Fmoc-L-glutamic acid y -t-butyl ester, 71989-18-9] ; MW 425.5; M.F. C24H27NO6 (25 g)
Catalog No. 50-845-555 Supplier AnaSpec Supplier No. AS20012
Fmoc-Glu(OtBu)-OH[N-a -Fmoc-L-glutamic acid y -t-butyl ester, 71989-18-9] ; MW 425.5; M.F. C24H27NO6 (25 g)