Learn More
Cayman Chemical Fmoc-Glu(OtBu)-OSu, Purity: ≥95%, CAS: 101214-22-6, 500 mg
SDP
Catalog No. 502990854
Encompass_Preferred
An amino acid-containing building block; used in the synthesis of bis- and tris-branched carboxyl group-terminal PEGylating reagents; has been used in the synthesis of N3S chelators
Catalog No. 50-299-0854 Supplier Cayman Chemical Supplier No. 30545500 MG
An amino acid-containing building block; used in the synthesis of bis- and tris-branched carboxyl group-terminal PEGylating reagents; has been used in the synthesis of N3S chelators