Learn More
Cayman Chemical Fmoc-Glu OtBu-OSu 5g
SDP
Catalog No. 502992287
Encompass_Preferred
An amino acid-containing building block; used in the synthesis of bis- and tris-branched carboxyl group-terminal PEGylating reagents; has been used in the synthesis of N3S chelators
Catalog No. 50-299-2287 Supplier Cayman Chemical Supplier No. 305455 G
An amino acid-containing building block; used in the synthesis of bis- and tris-branched carboxyl group-terminal PEGylating reagents; has been used in the synthesis of N3S chelators