Learn More
Sigma Aldrich Fine Chemicals Biosciences fmoc GluWang resin LL OAll
Catalog No. 502901046
Encompass
Useful resin for preparing head-to-tail cyclic peptides. Allyl group can be removed on the solid phase in the presence of standard t-butyl-based protecting groups by Pd-mediated allyl transfer.The product number for this product was previously 04-12-2093.To obtain a certificate of analysis (CoA) of a lot that begins with the letter A please select the option in the right hand menu Request a COA for Lots starting with A.
Catalog No. 50-290-1046 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 8561220005
Useful resin for preparing head-to-tail cyclic peptides. Allyl group can be removed on the solid phase in the presence of standard t-butyl-based protecting groups by Pd-mediated allyl transfer.The product number for this product was previously 04-12-2093.To obtain a certificate of analysis (CoA) of a lot that begins with the letter A please select the option in the right hand menu Request a COA for Lots starting with A.