Learn More
Bachem Fmoc-Gly-(Dmb)Gly-OH
SDP
Catalog No. 50312970
Encompass_Preferred
Fmoc-Gly-(Dmb)Gly-OH, 848861-65-4, C28H28N2O7, Mr 504.54, 1g. Introduction of the Dmb-moiety disrupts aggregated sequences during Fmoc-SPPS as efficiently as a pseudoproline residue. Dmb dipeptides couple smoothly, as, contrary to Hmb dipeptides, they cannot form oxazepinones. Dmb is removed during the final TFA cleavage in the presence of scavengers.
Catalog No. 50-312-970 Supplier Bachem Supplier No. 4062764.0001
Fmoc-Gly-(Dmb)Gly-OH, 848861-65-4, C28H28N2O7, Mr 504.54, 1g. Introduction of the Dmb-moiety disrupts aggregated sequences during Fmoc-SPPS as efficiently as a pseudoproline residue. Dmb dipeptides couple smoothly, as, contrary to Hmb dipeptides, they cannot form oxazepinones. Dmb is removed during the final TFA cleavage in the presence of scavengers.